Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Catalog No. 501885303
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM168. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Uterus, Blood vessels in all tissues, smooth muscle or leiomyosarcoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Crude human uterus extract. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 150kDa.Chromosome: 7q33. Gene Symbol: CALD1. Gene ID: 800. Ensemble: ENSG00000122786. Uniprot: NP_004333.
Catalog No. 50-188-5303 Supplier enQuire Bioreagents Supplier No. 800MSM1XP1
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM168. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Uterus, Blood vessels in all tissues, smooth muscle or leiomyosarcoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Crude human uterus extract. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 150kDa.Chromosome: 7q33. Gene Symbol: CALD1. Gene ID: 800. Ensemble: ENSG00000122786. Uniprot: NP_004333.