Learn More
Aviva Systems Biology Corp CALD1 Antibody - C-terminal region (ARP42684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CALD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CALD1. Alternate names: CDM, H-CAD, L-CAD, MGC21352, NAG22, HCAD, LCADApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58306