Learn More
Description
- This pocket chart makes teaching your students about the calendar and weather a breeze.
- Double-sided, full-color cards feature numbers, days, months, years, holidays, celebrations, seasons and weather conditions to encourage daily use.
- Storage pockets conveniently hold all of the cards.
- Included activity guide helps encourage a fun learning experience.
- Flame-retardant vinyl chart features grommets for easy hanging.
- Chart is designed for children ages 3 to 6.
- Learning Resources Calendar/Weather Pocket Chart
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Class |
| Product Type | Calendar |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
