Learn More
ATCC Calf Bovine Serum Iron Fortified (500 mL)
Supplier:  ATCC 302030
Encompass
Calf Bovine Serum is iron supplemented and triple filtered through 0.1 μm filters. Each lot of calf bovine serum is tested for sterility and for the ability to support the growth of cells using sequential growth curves. Calf bovine serum is manufactured from bovine blood collected in USDA-inspected abattoirs located in the United States. Serum has not come from cattle born, raised, shipped through, or slaughtered in countries where BSE is known to exist. This product has applications for cell culture, cell growth, and viability. Origin: Calf bovine serum is manufactured from bovine blood collected in USDA-inspected abattoirs located in the United States. Blood is collected from donor animals by venipuncture between the ages of 16 and 22 weeks. Calf bovine serum is supplemented with iron to off-set the high concentrations of transferrin produced by formula-fed animals prior to collection.
Catalog No. 50-238-4262