Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calf thymus DNA | 91080-16-9 | ≥97.0% | 50 MG
SDP

Catalog No. 5000345184
Encompass_Preferred

Calf thymus DNA is a high-quality double-stranded template DNA isolated from the thymus of male and female calves. It is used for research into the interaction between DNA and various agents.

  • High quality double-stranded template DNA
  • Isolated from the thymus of male and female calves
  • Used for research into DNA and agent interactions
  • Interacts with antiviral drug Lamivudine, studiable via emission, absorption, circular dichroism (CD), and viscosity techniques
  • Recommended to shake gently overnight at 0-4°C for complete dissolution to avoid shearing
  • Store at -20°C, protected from light
  • Soluble in H2O: ≥ 1 mg/mL

Catalog No. 50-003-45184 Supplier Medchemexpress LLC Supplier No. HY1095172MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calf thymus DNA is a high-quality double-stranded template DNA isolated from the thymus of male and female calves. It is used for research into the interaction between DNA and various agents.

  • High quality double-stranded template DNA
  • Isolated from the thymus of male and female calves
  • Used for research into DNA and agent interactions
  • Interacts with antiviral drug Lamivudine, studiable via emission, absorption, circular dichroism (CD), and viscosity techniques
  • Recommended to shake gently overnight at 0-4°C for complete dissolution to avoid shearing
  • Store at -20°C, protected from light
  • Soluble in H2O: ≥ 1 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.