Learn More
Sigma Aldrich Fine Chemicals Biosciences Calf Thymus Sonicated DNA
Catalog No. 502907582
Encompass
Genomic DNA sheared by sonication for use in hybridization experiments.Tested for absence of nuclease. Fragmented DNA with a size range from 200-6000 nucleotides. Purified by phenol extraction and freeze dried. Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 400 bp.
Catalog No. 50-290-7582 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE27456301
Genomic DNA sheared by sonication for use in hybridization experiments.Tested for absence of nuclease. Fragmented DNA with a size range from 200-6000 nucleotides. Purified by phenol extraction and freeze dried. Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 400 bp.