Learn More
SignaGen Laboratories CALFECTIN DNA REAGENT 1ML
Catalog No. 504791
Encompass
CalFectin Mammalian DNA Tranfection Reagent is a pre-optimized and refined version of traditional calcium phosphate transfection reagent. With a new chemistry, CalFectin Reagent contains much lower concentration of calcium, leading to much lower cytotoxicity with boosted transfection efficiency.
Catalog No. 50-479-1 Supplier SignaGen Laboratories Supplier No. SL100478
CalFectin Mammalian DNA Tranfection Reagent is a pre-optimized and refined version of traditional calcium phosphate transfection reagent. With a new chemistry, CalFectin Reagent contains much lower concentration of calcium, leading to much lower cytotoxicity with boosted transfection efficiency.