Learn More
SignaGen Laboratories CALFECTIN DNA REAGNT.1ML TRIAL
Catalog No. 504793
Encompass
CalFectin™ Mammalian DNA Tranfection Reagent is a pre-optimized and refined version of traditional calcium phosphate transfection reagent. With a new chemistry, CalFectin™ Reagent contains much lower concentration of calcium, leading to much lower cytotoxicity without sacrificing transfection efficiency. In addition, CalFectin™ Reagent is an effective method for the production of long-term stable and transient transfectants on most adherent cell lines. compared with liposome and polymer based transfection reagents, CalFectin™ Reagent shows distinguished features and much higher efficiency on HepG2, HEK293, CHO, Hela, COS, 3T3, LNcap, C6, PC12 and primary cultured cells, etc. CalFectin™ Reagent, 1.0 ml, is sufficient for ~666 transfections in 24 well plates or ~333 transfections in 6 well plates. Application High-efficiency stable or transient transfection of eukaryotic cells. - High efficient: Components optimized for high transfection efficiencies
Catalog No. 50-479-3 Supplier SignaGen Laboratories Supplier No. SL100478T
CalFectin™ Mammalian DNA Tranfection Reagent is a pre-optimized and refined version of traditional calcium phosphate transfection reagent. With a new chemistry, CalFectin™ Reagent contains much lower concentration of calcium, leading to much lower cytotoxicity without sacrificing transfection efficiency. In addition, CalFectin™ Reagent is an effective method for the production of long-term stable and transient transfectants on most adherent cell lines. compared with liposome and polymer based transfection reagents, CalFectin™ Reagent shows distinguished features and much higher efficiency on HepG2, HEK293, CHO, Hela, COS, 3T3, LNcap, C6, PC12 and primary cultured cells, etc. CalFectin™ Reagent, 1.0 ml, is sufficient for ~666 transfections in 24 well plates or ~333 transfections in 6 well plates. Application High-efficiency stable or transient transfection of eukaryotic cells. - High efficient: Components optimized for high transfection efficiencies