Learn More
Medchemexpress LLC CalFluor 488 Azide | 1798305-98-2 | C37H49N5O13S2 | 1 MG
SDP
Catalog No. 5000372948
Encompass_Preferred
CalFluor 488 Azide is a water-soluble fluorogenic azide probe that is activated by Cu-catalyzed or metal-free click reactions. It remains non-fluorescent until it reacts with alkynes, making it ideal for specific applications. The azide group allows for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules or ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Water-soluble fluorogenic azide probe
  • Activated by Cu-catalyzed or metal-free click reactions
  • Non-fluorescent until reaction with alkynes
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Capable of ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used for biological conjugation and label coupling
  • Categorized as a fluorescent dye and azide reactive dye for click chemistry
Catalog No. 50-003-72948 Supplier Medchemexpress LLC Supplier No. HY1517081MG
CalFluor 488 Azide is a water-soluble fluorogenic azide probe that is activated by Cu-catalyzed or metal-free click reactions. It remains non-fluorescent until it reacts with alkynes, making it ideal for specific applications. The azide group allows for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules or ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Water-soluble fluorogenic azide probe
  • Activated by Cu-catalyzed or metal-free click reactions
  • Non-fluorescent until reaction with alkynes
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Capable of ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used for biological conjugation and label coupling
  • Categorized as a fluorescent dye and azide reactive dye for click chemistry