Learn More
Medchemexpress LLC Calhex 231 hydrochloride | 2387505-78-2 | 99.8% | 443.41 g·mol⁻¹ | C25H28Cl2N2O | 1 ML
SDP
Catalog No. 5000244625
Encompass_Preferred
Calhex 231 hydrochloride is a research reagent that acts as a negative allosteric modulator of the calcium-sensing receptor (CaSR). It inhibits CaSR-mediated signaling (reported IC50 ≈ 0.39 μM) and is supplied in multiple formats including a ready-to-use 10 mM solution in DMSO for convenient dosing in cell-based and biochemical assays.
  • Negative allosteric modulator of CaSR for research applications.
  • Reported activity: IC50 ≈ 0.39 μM against CaSR-mediated responses.
  • Available as solid and as a pre-made 10 mM solution in DMSO (1 mL) for easy use.
  • High reported purity suitable for experimental use (reported 99.81%).
  • Provided with analytical documentation and safety data for laboratory compliance.
Catalog No. 50-002-44625 Supplier Medchemexpress LLC Supplier No. HY103320A10MM 1ML
Calhex 231 hydrochloride is a research reagent that acts as a negative allosteric modulator of the calcium-sensing receptor (CaSR). It inhibits CaSR-mediated signaling (reported IC50 ≈ 0.39 μM) and is supplied in multiple formats including a ready-to-use 10 mM solution in DMSO for convenient dosing in cell-based and biochemical assays.
  • Negative allosteric modulator of CaSR for research applications.
  • Reported activity: IC50 ≈ 0.39 μM against CaSR-mediated responses.
  • Available as solid and as a pre-made 10 mM solution in DMSO (1 mL) for easy use.
  • High reported purity suitable for experimental use (reported 99.81%).
  • Provided with analytical documentation and safety data for laboratory compliance.