Learn More
Medchemexpress LLC Calhex 231 hydrochloride | 2387505-78-2 | 99.8% | 443.41 g/mol | C25H28Cl2N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY103320A10MG
Encompass_Preferred
Calhex 231 hydrochloride is a research-grade small molecule that functions as a negative allosteric modulator of the calcium-sensing receptor (CaSR). It inhibits Ca2+-induced inositol phosphate accumulation (IC50 ≈ 0.39 μM) and is used in cellular studies related to traumatic hemorrhagic shock and diabetic cardiomyopathy.
  • Negative allosteric modulator of CaSR
  • Blocks Ca2+-induced inositol phosphate accumulation (IC50 ≈ 0.39 μM)
  • High purity (≈99.8%) suitable for in vitro research
  • White to off-white solid appearance
  • Stable when stored sealed and protected from moisture; recommended -20°C for solid storage
Catalog No. 50-002-45022