Learn More
Medchemexpress LLC Calhex 231 hydrochloride | 2387505-78-2 | MFCD18252543 | 99.8% | 443.41 g/mol | C25H28Cl2N2O | 50 MG
SDP
Catalog No. 5000245010
Encompass_Preferred
Calhex 231 hydrochloride is the hydrochloride salt of a small-molecule negative allosteric modulator of the calcium-sensing receptor (CaSR). It is supplied as a white to off-white solid with high reported purity for use in biochemical and cellular research.
  • Negative allosteric modulator of the calcium-sensing receptor (CaSR).
  • High reported purity (99.8%) suitable for research applications.
  • White to off-white solid, easy to handle and store.
  • Available in multiple package sizes and solution forms for flexible use.
  • Used as a tool compound in studies of CaSR signaling and related disease models.
Catalog No. 50-002-45010 Supplier Medchemexpress LLC Supplier No. HY103320A50MG
Calhex 231 hydrochloride is the hydrochloride salt of a small-molecule negative allosteric modulator of the calcium-sensing receptor (CaSR). It is supplied as a white to off-white solid with high reported purity for use in biochemical and cellular research.
  • Negative allosteric modulator of the calcium-sensing receptor (CaSR).
  • High reported purity (99.8%) suitable for research applications.
  • White to off-white solid, easy to handle and store.
  • Available in multiple package sizes and solution forms for flexible use.
  • Used as a tool compound in studies of CaSR signaling and related disease models.