Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calhex 231 hydrochloride | 2387505-78-2 | MFCD22580412 | 99.8% | 443.41 | C25H28Cl2N2O | 5 MG
SDP

Catalog No. 5000244775
Encompass_Preferred

Calhex 231 hydrochloride is the hydrochloride salt of Calhex 231, a negative allosteric modulator of the calcium-sensing receptor (CaSR) used in biochemical and cellular research. Supplied as a high-purity white to off-white solid, it is intended for in vitro studies and should be stored at -20°C in a sealed container.

  • Negative allosteric modulator of the CaSR.
  • CAS number 2387505-78-2.
  • Molecular formula C25H28Cl2N2O; molecular weight 443.41.
  • High purity (≈99.8%) suitable for research applications.
  • White to off-white solid; hydrochloride salt form.
  • Store sealed at -20°C and protect from moisture.
  • Used in studies of CaSR signaling, including trauma and cardiomyopathy models.

Catalog No. 50-002-44775 Supplier Medchemexpress LLC Supplier No. HY103320A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calhex 231 hydrochloride is the hydrochloride salt of Calhex 231, a negative allosteric modulator of the calcium-sensing receptor (CaSR) used in biochemical and cellular research. Supplied as a high-purity white to off-white solid, it is intended for in vitro studies and should be stored at -20°C in a sealed container.

  • Negative allosteric modulator of the CaSR.
  • CAS number 2387505-78-2.
  • Molecular formula C25H28Cl2N2O; molecular weight 443.41.
  • High purity (≈99.8%) suitable for research applications.
  • White to off-white solid; hydrochloride salt form.
  • Store sealed at -20°C and protect from moisture.
  • Used in studies of CaSR signaling, including trauma and cardiomyopathy models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.