Learn More
Medchemexpress LLC Calhex 231 hydrochloride | 2387505-78-2 | MFCD22580412 | 99.8% | 443.41 g·mol⁻¹ | C25H28Cl2N2O | 100 MG
SDP
Catalog No. 5000245093
Encompass_Preferred
Calhex 231 hydrochloride is a research-grade small molecule that functions as a negative allosteric modulator of the calcium-sensing receptor (CaSR), inhibiting CaSR-mediated inositol phosphate signaling. It is supplied as a solid, high-purity material intended for in vitro research applications.
  • Negative allosteric modulator of the calcium-sensing receptor (CaSR).
  • Reported IC50 ≈ 0.39 μM for inhibition of CaSR-mediated inositol phosphate signaling.
  • Solid powder with reported solubility in DMSO up to ~50 mM.
  • High purity (≈99.8%).
  • Intended for in vitro research use only.
Catalog No. 50-002-45093 Supplier Medchemexpress LLC Supplier No. HY103320A100MG
Calhex 231 hydrochloride is a research-grade small molecule that functions as a negative allosteric modulator of the calcium-sensing receptor (CaSR), inhibiting CaSR-mediated inositol phosphate signaling. It is supplied as a solid, high-purity material intended for in vitro research applications.
  • Negative allosteric modulator of the calcium-sensing receptor (CaSR).
  • Reported IC50 ≈ 0.39 μM for inhibition of CaSR-mediated inositol phosphate signaling.
  • Solid powder with reported solubility in DMSO up to ~50 mM.
  • High purity (≈99.8%).
  • Intended for in vitro research use only.