Learn More
VEOLIA WTS Calibration, 1,000-mg/L
Catalog No. NC0974294
Encompass
Set includes: (1) 250 mL bottle containing 1,000 mg C/L as KHP.
Catalog No. NC0974294 Supplier VEOLIA WTS Supplier No. STD 6841001
Set includes: (1) 250 mL bottle containing 1,000 mg C/L as KHP.