Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies Calibration Mix Majors, For AA and ICP-OES, 125 mL

Catalog No. NC3737666
Encompass

This calibration mix is designed for use in atomic absorption (AA) and inductively coupled plasma optical emission spectroscopy (ICP-OES) applications. The solution provides a precise standard for instrument calibration and validation.

  • Contains 5 analytes
  • Matrix composition of 5% HNO3
  • Concentration of Calcium (Ca) is 500 μg/mL
  • Supplied in a 125 mL volume

Catalog No. NC3737666 Supplier Agilent Technologies Supplier No. 6610030700
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This calibration mix is designed for use in atomic absorption (AA) and inductively coupled plasma optical emission spectroscopy (ICP-OES) applications. The solution provides a precise standard for instrument calibration and validation.

  • Contains 5 analytes
  • Matrix composition of 5% HNO3
  • Concentration of Calcium (Ca) is 500 μg/mL
  • Supplied in a 125 mL volume

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.