Learn More
Electron Microscopy Sciences Calibrated Loops 0.01 ml, 4 mm diameter, 70 mm length
SDP
Catalog No. 5024843
Encompass_Preferred
The 0.01 ml loops are calibrated to A.P.H.A. Standards and are made from 19 gauge B&S platinum wire with 5% rhodium.
Volume: 0.01 ml
Diameter: 4 mm
Length: 70 mm
Catalog No. 50-248-43 Supplier Electron Microscopy Sciences Supplier No. 6243020
The 0.01 ml loops are calibrated to A.P.H.A. Standards and are made from 19 gauge B&S platinum wire with 5% rhodium.
Volume: 0.01 ml
Diameter: 4 mm
Length: 70 mm