Learn More
Med Vet International Calibrated Microhematocrit Capillary Tubes, 75mm, J0542, 100/Tube, 10 Tubes/Box
Catalog No. 5000581780
Encompass_Preferred
These capillary tubes are designed for both safe blood collection as well as an accurate microhematocrit determination. Precision molded, they provide excellent sample retention during centrifugation. Tubes fill rapidly without affecting specimen integrity. Tube 1.15mm i.d. x 1.55mm o.d. 75mm length. Packaged in a protective snap cap vial. 100 tubes/vial/10 vials/cs
Catalog No. 50-005-81780 Supplier Med Vet International Supplier No. JOR542
These capillary tubes are designed for both safe blood collection as well as an accurate microhematocrit determination. Precision molded, they provide excellent sample retention during centrifugation. Tubes fill rapidly without affecting specimen integrity. Tube 1.15mm i.d. x 1.55mm o.d. 75mm length. Packaged in a protective snap cap vial. 100 tubes/vial/10 vials/cs