Learn More
METTLER TOLEDO™ Calibrated Thermometer
Catalog No. 13940070 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Calibrated Thermometer
Catalog No.
13-940-070 Calibrated Thermometer
1 options
Catalog No. 13-940-070 Supplier METTLER TOLEDO™ Supplier No. 11132685
Add to Cart
Add to Cart
Quick and Efficient Balance Conversion
  • Ensure Precise Temperatures
  • It is essential the water bath of the Density Kit is at the right temperature for correct density determination
  • Certified to Ensure Compliance
  • To help comply with quality guidelines, the certified thermometer comes with its own calibration certificate
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Water Bath of the Density Kit
For Use With (Application) Measuring density
Includes Calibration Certificate
Quantity 1
Type Thermometer