Learn More
Randox Clinical Chemistry Calibration Serum Level 3
SDP
Catalog No. NC9300478
Encompass_Preferred
Calibrator comprising 42 analytes in a single vial. This multi-analyte, third party calibrator is designed for use with a wide range of clinical chemistry platforms. Assayed, instrument, method and temperature specific values are supplied, ensuring accurate and reliable instrument calibration. • Lyophilised for enhanced stability • Human based serum • Stable to expiry date at 2ºC to 8ºC • Reconstituted stability of 7 days at 2ºC to 8ºC or 28 days at -20ºC
Catalog No. NC9300478 Supplier Randox Supplier No. CAL2351
Calibrator comprising 42 analytes in a single vial. This multi-analyte, third party calibrator is designed for use with a wide range of clinical chemistry platforms. Assayed, instrument, method and temperature specific values are supplied, ensuring accurate and reliable instrument calibration. • Lyophilised for enhanced stability • Human based serum • Stable to expiry date at 2ºC to 8ºC • Reconstituted stability of 7 days at 2ºC to 8ºC or 28 days at -20ºC