Learn More
Electron Microscopy Sciences Calibration Specimen 150-1D, Choose a mount
SDP
Catalog No. 501901661
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Has a nominal period (pitch) of 150 nm, one dimensional, fabricated on a transparent substrate (Aluminum lines on glass).
Mount: Choose a mount
Catalog No. 50-190-1661 Supplier Electron Microscopy Sciences Supplier No. 801251DX
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Has a nominal period (pitch) of 150 nm, one dimensional, fabricated on a transparent substrate (Aluminum lines on glass).
Mount: Choose a mount