Learn More
Electron Microscopy Sciences Calibration Specimen 300-2D, Steel disk for AFM
SDP
Catalog No. 501901674
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Array of Posts. Pitch 297. Al bumps on Si. Use contact or TappingMode.
Mount: AFM
Catalog No. 50-190-1674 Supplier Electron Microscopy Sciences Supplier No. 801232DAFM
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Array of Posts. Pitch 297. Al bumps on Si. Use contact or TappingMode.
Mount: AFM