Learn More
Electron Microscopy Sciences Calibration Specimen 300-1D, Choose a mount
SDP
Catalog No. 501901672
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Parallel Ridges. Pitch 288 W-coated Photoresist on Si Use contact or Tapping Mode.
Mount: Choose a mount
Catalog No. 50-190-1672 Supplier Electron Microscopy Sciences Supplier No. 801231DX
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Parallel Ridges. Pitch 288 W-coated Photoresist on Si Use contact or Tapping Mode.
Mount: Choose a mount