Learn More
Electron Microscopy Sciences Calibration Specimen 700-2D, Steel disk for AFM
SDP
Catalog No. 501901679
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Array of Posts. Pitch 700 W-coated Photoresist on Si. Use TappingMode.
Mount: AFM
Catalog No. 50-190-1679 Supplier Electron Microscopy Sciences Supplier No. 801222DAFM
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Array of Posts. Pitch 700 W-coated Photoresist on Si. Use TappingMode.
Mount: AFM