Learn More
Thermo Scientific™ Calibration Accessories for AQ7100 and AQ8100 Spectrophotometers
Catalog No. 15200949 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Orion AquaMate 7100/8100
Type:
Sample Holder for Film/Filter
Catalog No. For Use With (Equipment) Type
15-200-949 Orion AquaMate 7100/8100 Sample Holder for Film/Filter
1 options
Catalog No. 15-200-949 Supplier Thermo Scientific™ Supplier No. AQX1FLTRHDR
Add to Cart
Add to Cart
Check the performance and accuracy of your spectrophotometer right in your own laboratory using the Thermo Scientific™ SPECTRONIC Standards, and holder.
Calibration standards and film / filter holders ensure the accuracy of your AquaMate 7100 and 8100 Spectrophotometers. Check the performance and accuracy of your spectrophotometer right in your own laboratory using the Thermo Scientific™ SPECTRONIC Standards, and holder.
Sold separately, calibration standards and film / filter holders insure the accuracy of your AquaMate 7100 and 8100 Spectrophotometers.
Part number 840-253100 Includes:
Set of 7 glass pieces mounted in metal frames sized to fit standard square cuvette holders.
  • Four calibrated neutral density glass standards for checking photometric accuracy at nominal values of 0.3A, 0.5A, 1A and 1.5A◦Standards are calibrated at 440.0n, 465.0nm, 546.1nm, 590.0nm and 635.0nm
  • One calibrated standard for checking wavelength accuracy at nominal wavelengths on 400nm, 522nm and 777nm◦The symmetry of the peaks means that tests of wavelength accuracy are independent of the spectral bandwidth (between 1nm and 10nm) of the instrument being tested
  • filter for checking stray light in UV-Vis instruments at 220nm
  • 1 filter for checking stray light in Vis instruments at 400nm
  • Each set of SPECTRONIC Standards is individually tested and certified to be accurate, with values traceable to the National Institute of Standards and Technology (NIST) or other Primary Standards
  • Filters are supplied in a tough plastic storage container
Recalibration service is available from Thermo Fisher Scientific. Recommended recalibration interval is two years
Part Number AQX1FLTRHDR includes:
Film/Filter holder filters/lenses up to 50 mm long x 80 mm tall x 10 mm thick for Orion™ AquaMate 7100 & 8100 Spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications
Type Sample Holder for Film/Filter
For Use With (Equipment) Orion AquaMate 7100/8100
Holds Filters/lenses up to 50 mm long, 80 mm tall and 6 mm thick
Includes 1 Film / Filter Holder