Learn More
Cole-Parmer Techne FBCAL05D Liquid Calibration Bath with Cooling Coil; 5 L
Supplier:  Cole-Parmer 1611203
Encompass_Preferred
When temperature calibration is required the compact liquid baths offer excellent stability over the entire temperature range. These baths can be also be used for external circulation to maintain temperatures of samples in viscometers, photometers, refractometers, fermenters and other reaction vessels. All models offer high pump performance and exceptional thermal stability from -35°C to 250°C. The water baths are fully insulated on all sides and base and are fitted with a cooling coil for connection to a cold water supply for use at temperatures around ambient. The minimum temperature achievable is -35°C when a Flow Cooler is added to the system. Each bath is supplied complete with lid, drain tap, carry handles, a cooling coil (with bung) and hole to position a certified sensor. For Liquid Calibration Baths to operate, a thermoregulator is required. The thermoregulator must be purchased separately as models 01262-05 or 01262-07.
Catalog No. 50-196-1834