Learn More
Agilent Technologies CALIBRATION BLANK SOLUTI-500ML
Catalog No. NC2034584
Encompass
Calibration blank solution 5% HNO3 for ICP-OES, MP-AES, and AA, Certified Reference Material, 500 mL
Catalog No. NC2034584 Supplier Agilent Technologies Supplier No. 51907001
Calibration blank solution 5% HNO3 for ICP-OES, MP-AES, and AA, Certified Reference Material, 500 mL