Learn More
Gmp Labeling Inc Calibration By Status Label - Green
SDP
Catalog No. NC3482562
Encompass_Preferred
Roll labels, Thermal Transfer, Paper, GMP Labeling, 2" x 1", Green and White, Permanent Adhesive, 1000 labels/roll
Catalog No. NC3482562 Supplier Gmp Labeling Inc Supplier No. S066
Roll labels, Thermal Transfer, Paper, GMP Labeling, 2" x 1", Green and White, Permanent Adhesive, 1000 labels/roll