Learn More
Broadley James Corporation CALIBRATION CAP 12MM
Catalog No. NC0308506
Encompass
Calibration caps slide onto the sensing end of a probe providing an easy way to flush it with N2. This makes it easier to zero the sensor and check its responsiveness and calibration. Any tubing with a 6 mm (1/4″) i.d. attaches easily to the barbed end of the cap.
Catalog No. NC0308506 Supplier Broadley James Corporation Supplier No. AM9351
Calibration caps slide onto the sensing end of a probe providing an easy way to flush it with N2. This makes it easier to zero the sensor and check its responsiveness and calibration. Any tubing with a 6 mm (1/4″) i.d. attaches easily to the barbed end of the cap.