Learn More
BINDER™ Temperature Calibration Certificate (15 to 18 Measurement Points)
SDP
Catalog No. 08111620 Shop All BINDER Inc. Products
Change view
Click to view available options
Includes:
15 to 18 Measurement Points
Catalog No. Includes
08-111-620 15 to 18 Measurement Points
1 options
Catalog No. 08-111-620 Supplier BINDER™ Supplier No. 80121575
Add to Cart
Add to Cart
For Use With: FED 56, FED 115, FED 260, FED 400, FED 720
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Description Calibration certificate, temperature - temperature measurement incl. certificate, 15- 18 measuring points (at specified temperature)
For Use With (Equipment) BINDER Chambers FED 56, FED 115, FED 260, FED 400, FED 720
Type Temperature Calibration Certificate
For Use With (Application) Temperature Measurement
Includes 15 to 18 Measurement Points