Learn More
BINDER™ Calibration Certificate, Temperature and CO2
SDP
Catalog No. 5000217518 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Units Series CB, CBF
Catalog No. For Use With (Equipment)
50-002-17518 BINDER Units Series CB, CBF
1 options
Catalog No. 50-002-17518 Supplier BINDER™ Supplier No. 80121235
Provides official calibration certificates for temperature and CO2 to confirm that the units measured values are accurate and traceable to recognized standards
  • Calibration certificate, temperature and CO2 - for temperature and CO2
  • Temperature measurement in center of chamber
  • CO2 measurement performed using test gas at 37 °C and 5 % CO2
Specifications
For Use With (Equipment) BINDER Units Series CB, CBF
Weight (Metric) 1 kg