Learn More
Hach Company Calibration Cylinder Kit for Hach 1720C, 1720D, and 1720E Turbidimeters - includes calibration cylinder, precision pipette, and 500 mL 4000 NTU Formazin
Catalog No. 5000207003 Shop All Hach Company Products
Encompass
Calibration Cylinder Kit designed for use with Hach 1720-series turbidimeters. The kit provides the essential components to perform accurate turbidity calibration using a 4000 NTU formazin standard, enabling reliable verification and routine maintenance of instrument performance.
  • Provides a calibration cylinder for accurate sample measurement.
  • Includes a precision adjustable pipette (1.0-10.0 mL) for accurate dispensing.
  • Supplies 500 mL of 4000 NTU formazin calibration standard as a reference.
  • Helps ensure reliable, repeatable turbidity measurements.
  • Includes instructions to guide straightforward calibration procedures.
Catalog No. 50-002-07003 Supplier Hach Company Supplier No. 4415600
Calibration Cylinder Kit designed for use with Hach 1720-series turbidimeters. The kit provides the essential components to perform accurate turbidity calibration using a 4000 NTU formazin standard, enabling reliable verification and routine maintenance of instrument performance.
  • Provides a calibration cylinder for accurate sample measurement.
  • Includes a precision adjustable pipette (1.0-10.0 mL) for accurate dispensing.
  • Supplies 500 mL of 4000 NTU formazin calibration standard as a reference.
  • Helps ensure reliable, repeatable turbidity measurements.
  • Includes instructions to guide straightforward calibration procedures.