Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N064
Encompass_Preferred
The Grainger Approved Calibration Gas is a calibration gas cylinder that contains a 34L, 99.995% pure nitrogen gas blend. It comes equipped with a CGA 580 valve connection and is designed for instrument calibration and other analytical, industrial, and laboratory applications. This disposable cylinder has a capacity of 103 ft3 and a maximum allowable working pressure of 2265 psig at 70°F. The cylinder meets DOT 39 specifications and comes with a hydrostatic test date stamped on the shoulder.
Catalog No. 50-276-8361