Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N048
Encompass_Preferred
The 16N048 is a 16 oz. dead blow hammer by Malco Products. It has a non-marring urethane striking face to prevent surface damage. The hammer head contains shot to minimize rebound. It has an ergonomic, slip-resistant, dual material handle for comfort and grip. This hammer is ideal for assembling delicate materials or working in tight spaces where rebound is undesirable. It measures 13.5 inches long overall.
Catalog No. 50-276-5093