Learn More
Grainger Calibration Gas 116L Ammonia Nitrogen
Catalog No. 502764923
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing. It comes in a 34L cylinder with CGA 580 inlet connection. The gas purity is 99.998% nitrogen. The cylinder operating pressure is 1800 psig at 70°F. This calibration gas meets standards for use in calibration and testing of gas detection and analysis equipment. The product is available through Grainger.
Catalog No. 50-276-4923 Supplier Grainger Supplier No. 16N133
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing. It comes in a 34L cylinder with CGA 580 inlet connection. The gas purity is 99.998% nitrogen. The cylinder operating pressure is 1800 psig at 70°F. This calibration gas meets standards for use in calibration and testing of gas detection and analysis equipment. The product is available through Grainger.