Learn More
Grainger Calibration Gas 116L Carbon Dioxide Air
Supplier:  Grainger 16N076
Encompass_Preferred
The Gasco Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 34 liter aluminum cylinder contains 99.995% pure air balanced with nitrogen. It has a working pressure of 1800 psi with CGA-590 outlet connection. This calibration gas is recommended for calibrating and checking zero readings on gas detection instruments including combustible gas indicators, oxygen analyzers, and other air sensitive devices. The cylinder is refillable and includes a 5 year retest date. This is a high purity calibration gas used for accurate instrument calibration.
Catalog No. 50-276-8364