Learn More
Grainger Calibration Gas 116L Carbon Dioxide Air
Supplier:  Grainger 16N077
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing zero air. It has a capacity of 103L and is filled with 20.9% oxygen and 79.1% nitrogen. This high pressure cylinder has a CGA 590 outlet connection and is used for calibrating and checking the response of gas analyzers and monitoring equipment. The cylinder is manufactured by GASCO in the United States and is certified to meet NIST traceable standards. The cylinder must be secured in an upright position during storage and transport.
Catalog No. 50-276-5578