Learn More
Grainger Calibration Gas 116L Carbon Dioxide Air
Catalog No. 502768384
Encompass_Preferred
The Gasco Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. It has a capacity of 103L and contains 20.9% oxygen with the balance being nitrogen. The cylinder has a CGA-590 outlet connection and comes in an aluminum cylinder. The product has a 2 year recommended shelf life and is manufactured in the USA. It can be used for calibrating and bump testing gas detectors to ensure proper operation and measurement. The cylinder is DOT approved for shipment.
Catalog No. 50-276-8384 Supplier Grainger Supplier No. 16N073
The Gasco Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. It has a capacity of 103L and contains 20.9% oxygen with the balance being nitrogen. The cylinder has a CGA-590 outlet connection and comes in an aluminum cylinder. The product has a 2 year recommended shelf life and is manufactured in the USA. It can be used for calibrating and bump testing gas detectors to ensure proper operation and measurement. The cylinder is DOT approved for shipment.