Learn More
Grainger Calibration Gas 116L Carbon Monoxide Air
Catalog No. 502768509
Encompass_Preferred
The Grainger 116L Calibration Gas Carbon Monoxide Air is a 116 liter cylinder of calibration gas containing 10 parts per million carbon monoxide in air. This gas mixture is used for calibration and bump testing of carbon monoxide gas detectors to ensure proper operation and response. The cylinder has a CGA-590 outlet connection and comes equipped with a carrying strap for portability. The gas mixture is certified to NIST traceable standards and has a shelf life of 18 months. This product can be used for calibration of carbon monoxide sensors in gas detection equipment.
Catalog No. 50-276-8509 Supplier Grainger Supplier No. 16N080
The Grainger 116L Calibration Gas Carbon Monoxide Air is a 116 liter cylinder of calibration gas containing 10 parts per million carbon monoxide in air. This gas mixture is used for calibration and bump testing of carbon monoxide gas detectors to ensure proper operation and response. The cylinder has a CGA-590 outlet connection and comes equipped with a carrying strap for portability. The gas mixture is certified to NIST traceable standards and has a shelf life of 18 months. This product can be used for calibration of carbon monoxide sensors in gas detection equipment.