Learn More
Grainger Calibration Gas 116L Chlorine Nitrogen
Catalog No. 502767120
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 116 liter cylinder with a CGA 580 outlet connection. The cylinder operating pressure is 2000 psig. This calibration gas meets standards for use in calibration and testing of gas detection and analysis equipment. The product number for ordering is 16N141. This is a high quality nitrogen gas for analytical, scientific, and calibration applications.
Catalog No. 50-276-7120 Supplier Grainger Supplier No. 16N141
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 116 liter cylinder with a CGA 580 outlet connection. The cylinder operating pressure is 2000 psig. This calibration gas meets standards for use in calibration and testing of gas detection and analysis equipment. The product number for ordering is 16N141. This is a high quality nitrogen gas for analytical, scientific, and calibration applications.