Learn More
Grainger Calibration Gas 116L Chlorine Nitrogen
Catalog No. 502767720
Encompass_Preferred
The Grainger 10ppm Chlorine Cylinder Calibration Gas is a high-precision gas mixture used for calibration and bump testing of chlorine gas detectors. It comes in a 34L aluminum cylinder with CGA-350 outlet connection. The gas mixture contains 10 parts per million chlorine with balance nitrogen. It has a 2 year recommended shelf life. This calibration gas is suitable for chlorine leak detection in water treatment, chemical processing, and other industrial applications. It enables compliance with calibration requirements and ensures chlorine gas detectors function properly.
Catalog No. 50-276-7720 Supplier Grainger Supplier No. 16N139
The Grainger 10ppm Chlorine Cylinder Calibration Gas is a high-precision gas mixture used for calibration and bump testing of chlorine gas detectors. It comes in a 34L aluminum cylinder with CGA-350 outlet connection. The gas mixture contains 10 parts per million chlorine with balance nitrogen. It has a 2 year recommended shelf life. This calibration gas is suitable for chlorine leak detection in water treatment, chemical processing, and other industrial applications. It enables compliance with calibration requirements and ensures chlorine gas detectors function properly.