Learn More
Grainger Calibration Gas 116L Methane Air
Catalog No. 502766136
Encompass_Preferred
The Gasco Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 16 liter aluminum cylinder contains 99.9995% pure air certified to NIST traceable standards. The cylinder has a CGA-590 outlet with a brass valve and is refillable. This calibration gas is suitable for calibrating and bump testing gas detectors used for monitoring oxygen levels. With a 2 year shelf life, this cylinder allows for reliable instrument calibration. The 16N117 model includes a carrying case for portability.
Catalog No. 50-276-6136 Supplier Grainger Supplier No. 16N117
The Gasco Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 16 liter aluminum cylinder contains 99.9995% pure air certified to NIST traceable standards. The cylinder has a CGA-590 outlet with a brass valve and is refillable. This calibration gas is suitable for calibrating and bump testing gas detectors used for monitoring oxygen levels. With a 2 year shelf life, this cylinder allows for reliable instrument calibration. The 16N117 model includes a carrying case for portability.