Learn More
Grainger Calibration Gas 116L Propane Air
Catalog No. 502765810
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing 20.9% oxygen gas balanced with nitrogen. It comes in a 103 liter aluminum cylinder with a CGA 590 outlet connection. The cylinder operating pressure is 1800 psi. This calibration gas is used for checking, calibrating, and bump testing gas detection instruments to ensure they are functioning properly and accurately detecting oxygen levels. The cylinder is refillable and is built to DOT 3AA 2015 specifications. The product has a 12 month shelf life and is manufactured in the USA. This calibration gas aids in compliance with OSHA safety regulations.
Catalog No. 50-276-5810 Supplier Grainger Supplier No. 16N130
The GASCO Calibration Gas - Air is a calibration gas cylinder containing 20.9% oxygen gas balanced with nitrogen. It comes in a 103 liter aluminum cylinder with a CGA 590 outlet connection. The cylinder operating pressure is 1800 psi. This calibration gas is used for checking, calibrating, and bump testing gas detection instruments to ensure they are functioning properly and accurately detecting oxygen levels. The cylinder is refillable and is built to DOT 3AA 2015 specifications. The product has a 12 month shelf life and is manufactured in the USA. This calibration gas aids in compliance with OSHA safety regulations.