Learn More
Grainger Calibration Gas 116L Pure Hydrogen
Supplier:  Grainger 16N106
Encompass_Preferred
The Grainger Approved Calibration Gas is a 16 oz disposable cylinder of calibration gas used for calibrating gas detection instruments. It contains 50 ppm hydrogen sulfide gas balanced with nitrogen. The cylinder has a brass CGA-350 outlet valve and calibration instructions are included. This calibration gas has a 12 month shelf life and is DOT approved for transport of hazardous materials. It can be used to calibrate and bump test H2S gas detectors to ensure proper functionality and calibration. This is a single-use disposable cylinder that cannot be refilled.
Catalog No. 50-276-7362