Learn More
Grainger Calibration Gas 116L Pure Nitrogen
Catalog No. 502765625
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing 20.9% oxygen gas balanced with nitrogen. It comes in a 103 liter aluminum cylinder with a CGA 590 outlet connection. The cylinder operating pressure is 1800 psi. This calibration gas is used for checking, calibrating, and bump testing gas detection instruments to ensure they are functioning properly and accurately detecting oxygen levels. The cylinder is refillable and is built to DOT 3AA 2015 specifications. The product has a 12 month shelf life and is available through Grainger.
Catalog No. 50-276-5625 Supplier Grainger Supplier No. 16N119
The GASCO Calibration Gas - Air is a calibration gas cylinder containing 20.9% oxygen gas balanced with nitrogen. It comes in a 103 liter aluminum cylinder with a CGA 590 outlet connection. The cylinder operating pressure is 1800 psi. This calibration gas is used for checking, calibrating, and bump testing gas detection instruments to ensure they are functioning properly and accurately detecting oxygen levels. The cylinder is refillable and is built to DOT 3AA 2015 specifications. The product has a 12 month shelf life and is available through Grainger.