Learn More
Grainger Calibration Gas 116L Sulfur Dioxide Air
Catalog No. 502767430
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing zero air. It has a capacity of 103L and is filled with 20.9% oxygen and 79.1% nitrogen. This gas mixture is used for calibration and bump testing of gas detection instruments to ensure proper operation and measurement. The cylinder has a CGA-590 outlet connection and comes in a lightweight aluminum cylinder for portability. The product has a 2 year shelf life and is manufactured in the USA under strict quality control standards. It can be used for calibration of combustible gas detectors, multi-gas instruments, and other air sensitive devices.
Catalog No. 50-276-7430 Supplier Grainger Supplier No. 16N179
The GASCO Calibration Gas - Air is a calibration gas cylinder containing zero air. It has a capacity of 103L and is filled with 20.9% oxygen and 79.1% nitrogen. This gas mixture is used for calibration and bump testing of gas detection instruments to ensure proper operation and measurement. The cylinder has a CGA-590 outlet connection and comes in a lightweight aluminum cylinder for portability. The product has a 2 year shelf life and is manufactured in the USA under strict quality control standards. It can be used for calibration of combustible gas detectors, multi-gas instruments, and other air sensitive devices.