Learn More
Electron Microscopy Sciences Calibration Slide for Hardness Testers Brinell
SDP
Catalog No. 501901562
Encompass_Preferred
Reflected light calibration slide with circle shapes and scales for calibrating Brinell hardness testers. The PS26 which has a series of circles, to represent the ball indentation shape, of varying size and x-y scales. It is a precision marked chrome deposition image on a glass slide. Each of the markings on the slide is clearly identified with its size. This product is supplied in a polished wooden case.
PS26 Pattern Detail: Circles: (Diameter, mm's) 5.0, 2.5, 1.0, 0.7, 0.3
Scales: Horizontal and vertical, 2 mm in 0.1 mm divisions
Catalog No. 50-190-1562 Supplier Electron Microscopy Sciences Supplier No. 6804915
Reflected light calibration slide with circle shapes and scales for calibrating Brinell hardness testers. The PS26 which has a series of circles, to represent the ball indentation shape, of varying size and x-y scales. It is a precision marked chrome deposition image on a glass slide. Each of the markings on the slide is clearly identified with its size. This product is supplied in a polished wooden case.
PS26 Pattern Detail: Circles: (Diameter, mm's) 5.0, 2.5, 1.0, 0.7, 0.3
Scales: Horizontal and vertical, 2 mm in 0.1 mm divisions