Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO CALIBRATION HEATER 5 W ALLOY

Catalog No. 502741613 Shop All METTLER TOLEDO Products
Encompass

Calibration heater 5W HC (Hockey type)

Catalog No. 50-274-1613 Supplier METTLER TOLEDO Supplier No. 51160179
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Calibration heater 5W HC (Hockey type)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.