Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO CALIBRATION HEATER 5W 6/230MM

Catalog No. 502741515 Shop All METTLER TOLEDO Products
Encompass

Calibration heater 5 W, glass

Catalog No. 50-274-1515 Supplier METTLER TOLEDO Supplier No. 51162457

missing translation for 'validMainframeMsg'

Calibration heater 5 W, glass

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.