Learn More
VAISALA INC Calibration Salt, LiCl, 11% RH | 7447-41-8 | MFCD00011078
Catalog No. NC1142646
Encompass_Preferred
Calibration Salt, LiCl, 11% RH | MW: 42.394 | 7447-41-8 | MFCD00011078
Catalog No. NC1142646 Supplier VAISALA INC Supplier No. 19729HM
Calibration Salt, LiCl, 11% RH | MW: 42.394 | 7447-41-8 | MFCD00011078